Assay Details
| Gene Symbol: PPIE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678654.3 | XP_005678711.1 | 9 - 10 | 1234 | 57 | |
| XM_005678655.2 | XP_005678712.1 | 9 - 10 | 870 | 57 | ||
| GenBank mRNA | KA353090.1 | - | 2 - 3 | 237 | 57 | |
More Information
Set Membership: |
|

PPIE
Genomic Map