Assay Details
Gene Symbol: LOC102190507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682628.2 | XP_005682685.2 | 4 - 5 | 323 | 77 | |
XM_013965276.1 | XP_013820730.1 | 4 - 5 | 335 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
