Assay Details
| Gene Symbol: LOC102186703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688996.2 | XP_005689053.1 | 2 - 3 | 454 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102186703| Gene Symbol: LOC102186703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688996.2 | XP_005689053.1 | 2 - 3 | 454 | 78 | |
Set Membership: |
|