Assay Details
| Gene Symbol: SLC25A19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694142.3 | XP_005694199.1 | 3 - 4 | 313 | 57 | |
| XM_013972239.2 | XP_013827693.1 | 3 - 4 | 314 | 57 | ||
| GenBank mRNA | JO593132.2 | - | 4 - 5 | 1450 | 57 | |
| JO593133.2 | - | 5 - 6 | 1621 | 57 | ||
More Information
Set Membership: |
|

SLC25A19
Genomic Map