Assay Details
| Gene Symbol: ARL6IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697528.3 | XP_005697585.1 | 5 - 6 | 691 | 67 | |
| GenBank mRNA | JO429792.2 | - | 5 - 6 | 604 | 67 | |
| JO579869.1 | - | 1 - 2 | 1564 | 67 | ||
More Information
Set Membership: |
|

ARL6IP1
Genomic Map