Assay Details
| Gene Symbol: AQP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697024.2 | XP_005697081.1 | 3 - 4 | 737 | 61 | |
| XM_005697025.3 | XP_005697082.1 | 3 - 4 | 837 | 61 | ||
| XM_005697026.3 | XP_005697083.1 | 3 - 4 | 1083 | 61 | ||
| GenBank mRNA | JO633330.2 | - | 2 - 3 | 590 | 61 | |
More Information
Set Membership: |
|

AQP4
Genomic Map