Assay Details
| Gene Symbol: ICAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682354.3 | XP_005682411.2 | 2 - 3 | 387 | 89 | |
| GenBank mRNA | JO559125.2 | - | 2 - 3 | 407 | 89 | |
More Information
Set Membership: |
|

ICAM1
Genomic Map