Assay Details
| Gene Symbol: RABAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062502.1 | XP_017917991.1 | 2 - 3 | 406 | 58 | |
| GenBank mRNA | JO581518.1 | - | 2 - 3 | 529 | 58 | |
More Information
Set Membership: |
|

RABAC1
Genomic Map