Assay Details
| Gene Symbol: ID3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055625.1 | XP_017911114.1 | 2 - 3 | 589 | 99 | |
| GenBank mRNA | JO517612.2 | - | 2 - 3 | 193 | 99 | |
| JO563986.2 | - | 2 - 3 | 487 | 99 | ||
More Information
Set Membership: |
|

ID3
Genomic Map