Assay Details
| Gene Symbol: LOC102185866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065112.1 | XP_017920601.1 | 12 - 13 | 1641 | 66 | |
| XM_018065113.1 | XP_017920602.1 | 11 - 12 | 1808 | 66 | ||
| XM_018065114.1 | XP_017920603.1 | 11 - 12 | 2035 | 66 | ||
| GenBank mRNA | JN688205.1 | - | 12 - 13 | 1624 | 66 | |
More Information
Set Membership: |
|

LOC102185866
Genomic Map