Assay Details
| Gene Symbol: SCD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049650.1 | XP_017905139.1 | 3 - 4 | 914 | 74 | |
| GenBank mRNA | JO633672.2 | - | 3 - 4 | 503 | 74 | |
More Information
Set Membership: |
|

SCD5
Genomic Map