Assay Details
| Gene Symbol: LOC106504024 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044656.1 | XP_017900145.1 | 2430 | 70 | ||
| Gene Symbol: HTT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049859.1 | XP_017905348.1 | 55 - 56 | 7912 | 70 | |
| XM_018049860.1 | XP_017905349.1 | 55 - 56 | 7572 | 70 | ||
| XM_018049861.1 | XP_017905350.1 | 54 - 55 | 7864 | 70 | ||
| XM_018049862.1 | XP_017905351.1 | 53 - 54 | 7780 | 70 | ||
More Information
Set Membership: |
|

LOC106504024
Genomic Map