Assay Details
| Gene Symbol: NECTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064224.1 | XP_017919713.1 | 3 - 4 | 963 | 87 | |
| XM_018064281.1 | XP_017919770.1 | 3 - 4 | 963 | 87 | ||
| GenBank mRNA | JO361414.2 | - | 1 - 2 | 274 | 87 | |
| JO397501.2 | - | 3 - 4 | 1040 | 87 | ||
| JO444202.2 | - | 3 - 4 | 702 | 87 | ||
| JO444979.2 | - | 3 - 4 | 702 | 87 | ||
| KA340304.1 | - | 3 - 4 | 731 | 87 | ||
More Information
Set Membership: |
|

NECTIN3
Genomic Map