Assay Details
| Gene Symbol: NECTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064281.1 | XP_017919770.1 | 8 - 9 | 1514 | 84 | |
| GenBank mRNA | JO398764.2 | - | 1 - 2 | 242 | 84 | |
| JO456863.2 | - | 3 - 4 | 286 | 84 | ||
More Information
Set Membership: |
|

NECTIN3
Genomic Map