Assay Details
| Gene Symbol: CPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675465.3 | XP_005675522.2 | 9 - 10 | 993 | 60 | |
| GenBank mRNA | KA335269.1 | - | 2 - 3 | 618 | 60 | |
More Information
Set Membership: |
|

CPA3
Genomic Map