Assay Details
| Gene Symbol: PLPPR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678058.3 | XP_005678115.1 | 1 - 2 | 440 | 82 | |
| XM_005678059.3 | XP_005678116.1 | 1 - 2 | 418 | 82 | ||
More Information
Set Membership: |
|

PLPPR5
Genomic Map