Assay Details
| Gene Symbol: PLPPR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678058.3 | XP_005678115.1 | 3 - 4 | 824 | 70 | |
| XM_005678059.3 | XP_005678116.1 | 3 - 4 | 802 | 70 | ||
More Information
Set Membership: |
|

PLPPR5
Genomic Map