Assay Details
| Gene Symbol: SVBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042682.1 | XP_017898171.1 | 2 - 3 | 217 | 64 | |
| XM_018042685.1 | XP_017898174.1 | 2 - 3 | 496 | 64 | ||
| XM_018042695.1 | XP_017898184.1 | 3 - 4 | 343 | 64 | ||
| XM_018042701.1 | XP_017898190.1 | 3 - 4 | 625 | 64 | ||
| XM_018042710.1 | XP_017898199.1 | 2 - 3 | 227 | 64 | ||
| XM_018042719.1 | XP_017898208.1 | 2 - 3 | 242 | 64 | ||
| XM_018042729.1 | XP_017898218.1 | 2 - 3 | 238 | 64 | ||
| XM_018042733.1 | XP_017898222.1 | 2 - 3 | 502 | 64 | ||
More Information
Set Membership: |
|

SVBP
Genomic Map