Assay Details
| Gene Symbol: MARCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050994.1 | XP_017906483.1 | 2 - 3 | 363 | 75 | |
| GenBank mRNA | JT634796.1 | - | 3 - 4 | 412 | 75 | |
| JT664150.1 | - | 2 - 3 | 345 | 75 | ||
More Information
Set Membership: |
|

MARCH2
Genomic Map