Assay Details
| Gene Symbol: LOC102189116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682625.3 | XP_005682682.1 | 10 - 11 | 1440 | 60 | |
| XM_013965331.2 | XP_013820785.1 | 8 - 9 | 1172 | 60 | ||
| XM_013965333.2 | XP_013820787.1 | 8 - 9 | 1172 | 60 | ||
| XM_018050726.1 | XP_017906215.1 | 10 - 11 | 1439 | 60 | ||
| XM_018050727.1 | XP_017906216.1 | 10 - 11 | 1509 | 60 | ||
| XM_018050728.1 | XP_017906217.1 | 11 - 12 | 1544 | 60 | ||
| XM_018050729.1 | XP_017906218.1 | 10 - 11 | 1430 | 60 | ||
| XM_018050730.1 | XP_017906219.1 | 10 - 11 | 1439 | 60 | ||
| GenBank mRNA | JO427600.1 | - | 13 - 14 | 1846 | 60 | |
More Information
Set Membership: |
|

LOC102189116
Genomic Map