Assay Details
| Gene Symbol: LOC102189116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682625.3 | XP_005682682.1 | 12 - 13 | 1587 | 67 | |
| XM_018050726.1 | XP_017906215.1 | 12 - 13 | 1586 | 67 | ||
| XM_018050727.1 | XP_017906216.1 | 12 - 13 | 1656 | 67 | ||
| XM_018050728.1 | XP_017906217.1 | 13 - 14 | 1691 | 67 | ||
| XM_018050729.1 | XP_017906218.1 | 12 - 13 | 1577 | 67 | ||
| GenBank mRNA | JO427600.1 | - | 15 - 16 | 1993 | 67 | |
More Information
Set Membership: |
|

LOC102189116
Genomic Map