Assay Details
| Gene Symbol: CNOT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683212.3 | XP_005683269.1 | 6 - 7 | 1086 | 79 | |
| XM_005683213.2 | XP_005683270.1 | 6 - 7 | 957 | 79 | ||
| XM_013965718.2 | XP_013821172.1 | 4 - 5 | 539 | 79 | ||
| GenBank mRNA | JO433114.2 | - | 1 - 2 | 706 | 79 | |
| JO576908.1 | - | 6 - 7 | 1087 | 79 | ||
More Information
Set Membership: |
|

CNOT8
Genomic Map