Assay Details
| Gene Symbol: FAXDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051641.1 | XP_017907130.1 | 7 - 8 | 1360 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAXDC2| Gene Symbol: FAXDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051641.1 | XP_017907130.1 | 7 - 8 | 1360 | 59 | |
Set Membership: |
|