Assay Details
| Gene Symbol: DOP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052919.1 | XP_017908408.1 | 20 - 21 | 5316 | 139 | |
| XM_018052920.1 | XP_017908409.1 | 20 - 21 | 5282 | 139 | ||
| XM_018052921.1 | XP_017908410.1 | 19 - 20 | 5252 | 139 | ||
| XM_018052922.1 | XP_017908411.1 | 19 - 20 | 5118 | 139 | ||
| XM_018052923.1 | XP_017908412.1 | 20 - 21 | 5282 | 139 | ||
| XM_018052924.1 | XP_017908413.1 | 20 - 21 | 5282 | 139 | ||
| GenBank mRNA | JT661234.1 | - | 21 - 22 | 5068 | 135 | |
More Information
Set Membership: |
|

DOP1A
Genomic Map