Assay Details
| Gene Symbol: DOP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052919.1 | XP_017908408.1 | 37 - 38 | 7294 | 77 | |
| XM_018052920.1 | XP_017908409.1 | 37 - 38 | 7260 | 77 | ||
| XM_018052921.1 | XP_017908410.1 | 36 - 37 | 7230 | 77 | ||
| XM_018052924.1 | XP_017908413.1 | 37 - 38 | 7142 | 77 | ||
| GenBank mRNA | JT661234.1 | - | 40 - 41 | 7018 | 77 | |
More Information
Set Membership: |
|

DOP1A
Genomic Map