Assay Details
| Gene Symbol: DOP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052919.1 | XP_017908408.1 | 38 - 39 | 7351 | 71 | |
| XM_018052920.1 | XP_017908409.1 | 38 - 39 | 7317 | 71 | ||
| XM_018052921.1 | XP_017908410.1 | 37 - 38 | 7287 | 71 | ||
More Information
Set Membership: |
|

DOP1A
Genomic Map