Assay Details
| Gene Symbol: GLCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053588.1 | XP_017909077.1 | 3 - 4 | 1015 | 79 | |
| XM_018053589.1 | XP_017909078.1 | 2 - 3 | 419 | 79 | ||
| GenBank mRNA | JO411270.2 | - | 3 - 4 | 1040 | 79 | |
| JT716294.1 | - | 3 - 4 | 1006 | 79 | ||
More Information
Set Membership: |
|

GLCE
Genomic Map