Assay Details
| Gene Symbol: LOC102169480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685235.3 | XP_005685292.3 | 2 - 3 | 503 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102169480| Gene Symbol: LOC102169480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685235.3 | XP_005685292.3 | 2 - 3 | 503 | 86 | |
Set Membership: |
|