Assay Details
| Gene Symbol: WBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686330.3 | XP_005686387.1 | 3 - 4 | 439 | 74 | |
| XM_005686332.3 | XP_005686389.1 | 2 - 3 | 371 | 74 | ||
| GenBank mRNA | JO587300.2 | - | 1 - 2 | 57 | 74 | |
More Information
Set Membership: |
|

WBP1
Genomic Map