Assay Details
| Gene Symbol: GPATCH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686484.3 | XP_005686541.2 | 6 - 7 | 705 | 61 | |
| XM_018055125.1 | XP_017910614.1 | 7 - 8 | 822 | 61 | ||
| GenBank mRNA | JO588067.2 | - | 3 - 4 | 1385 | 61 | |
| JO588068.2 | - | 3 - 4 | 1385 | 61 | ||
More Information
Set Membership: |
|

GPATCH11
Genomic Map