Assay Details
| Gene Symbol: TOR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687160.3 | XP_005687217.2 | 2 - 3 | 557 | 55 | |
| XM_018055818.1 | XP_017911307.1 | 2 - 3 | 557 | 55 | ||
| GenBank mRNA | JT680714.1 | - | 5 - 6 | 1135 | 55 | |
More Information
Set Membership: |
|

TOR2A
Genomic Map