Assay Details
| Gene Symbol: NOP56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057443.1 | XP_017912932.1 | 7 - 8 | 1030 | 59 | |
| GenBank mRNA | JO610516.2 | - | 5 - 6 | 1151 | 59 | |
More Information
Set Membership: |
|

NOP56
Genomic Map