Assay Details
| Gene Symbol: CDH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692102.3 | XP_005692159.1 | 3 - 4 | 647 | 56 | |
| XM_018062082.1 | XP_017917571.1 | 3 - 4 | 647 | 56 | ||
| GenBank mRNA | JO427362.1 | - | 3 - 4 | 677 | 56 | |
| KA313650.1 | - | 3 - 4 | 674 | 56 | ||
More Information
Set Membership: |
|

CDH5
Genomic Map