Assay Details
| Gene Symbol: DHODH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692197.3 | XP_005692254.2 | 7 - 8 | 1042 | 64 | |
| XM_005692201.3 | XP_005692258.1 | 6 - 7 | 811 | 64 | ||
| XM_013971133.2 | XP_013826587.2 | 8 - 9 | 1138 | 64 | ||
| XM_013971134.2 | XP_013826588.2 | 8 - 9 | 1109 | 64 | ||
| GenBank mRNA | JO414849.2 | - | 2 - 3 | 616 | 64 | |
More Information
Set Membership: |
|

DHODH
Genomic Map