Assay Details
| Gene Symbol: ZNF793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013971210.2 | XP_013826664.1 | 4 - 5 | 410 | 76 | |
| XM_013971211.2 | XP_013826665.1 | 4 - 5 | 407 | 76 | ||
| XM_013971212.2 | XP_013826666.1 | 2 - 3 | 164 | 76 | ||
| GenBank mRNA | KA314555.1 | - | 2 - 3 | 312 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013971212.2 transcripts.
Set Membership: |
|

ZNF793
Genomic Map