Assay Details
| Gene Symbol: YIF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062327.1 | XP_017917816.1 | 7 - 8 | 948 | 59 | |
| XM_018062328.1 | XP_017917817.1 | 7 - 8 | 936 | 59 | ||
| GenBank mRNA | KA335378.1 | - | 1 - 2 | 368 | 59 | |
More Information
Set Membership: |
|

YIF1B
Genomic Map