Assay Details
| Gene Symbol: RANBP3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065805.1 | XP_017921294.1 | 10 - 11 | 1361 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RANBP3L| Gene Symbol: RANBP3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065805.1 | XP_017921294.1 | 10 - 11 | 1361 | 73 | |
Set Membership: |
|