Assay Details
| Gene Symbol: LOC102174081 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039365.1 | XP_017894854.1 | 12 - 13 | 1578 | 88 | |
| GenBank mRNA | KA313924.1 | - | 2 - 3 | 318 | 88 | |
More Information
Set Membership: |
|

LOC102174081
Genomic Map