Assay Details
| Gene Symbol: LOC102174081 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039365.1 | XP_017894854.1 | 14 - 15 | 1907 | 71 | |
| GenBank mRNA | JO621783.2 | - | 1 - 2 | 143 | 71 | |
More Information
Set Membership: |
|

LOC102174081
Genomic Map