Assay Details
| Gene Symbol: CIAO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040437.1 | XP_017895926.1 | 7 - 8 | 1225 | 58 | |
| XM_018040438.1 | XP_017895927.1 | 7 - 8 | 1473 | 58 | ||
| XM_018040439.1 | XP_017895928.1 | 7 - 8 | 1271 | 58 | ||
| XM_018040441.1 | XP_017895930.1 | 7 - 8 | 1186 | 58 | ||
| GenBank mRNA | JO641454.2 | - | 4 - 5 | 1258 | 58 | |
More Information
Set Membership: |
|

CIAO3
Genomic Map