Assay Details
| Gene Symbol: TNFRSF12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040800.1 | XP_017896289.1 | 3 - 4 | 407 | 86 | |
| GenBank mRNA | JO571485.2 | - | 3 - 4 | 360 | 86 | |
| JT670627.1 | - | 3 - 4 | 321 | 86 | ||
More Information
Set Membership: |
|

TNFRSF12A
Genomic Map