Assay Details
| Gene Symbol: HIRIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697676.3 | XP_005697733.2 | 5 - 6 | 1463 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIRIP3| Gene Symbol: HIRIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697676.3 | XP_005697733.2 | 5 - 6 | 1463 | 61 | |
Set Membership: |
|