Assay Details
Gene Symbol: R3HCC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698308.3 | XP_005698365.2 | 4 - 5 | 2023 | 127 | |
XM_018041558.1 | XP_017897047.1 | 3 - 4 | 1974 | 127 | ||
XM_018041559.1 | XP_017897048.1 | 5 - 6 | 1399 | 127 | ||
GenBank mRNA | JO612893.2 | - | 6 - 7 | 2130 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
