Assay Details
| Gene Symbol: PDLIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698381.3 | XP_005698438.3 | 5 - 6 | 874 | 94 | |
| GenBank mRNA | JO611392.2 | - | 5 - 6 | 835 | 94 | |
More Information
Set Membership: |
|

PDLIM1
Genomic Map