Assay Details
| Gene Symbol: DCTD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698704.3 | XP_005698761.1 | 5 - 6 | 547 | 68 | |
| XM_018042125.1 | XP_017897614.1 | 5 - 6 | 538 | 68 | ||
| GenBank mRNA | JI861759.1 | - | 4 - 5 | 460 | 68 | |
More Information
Set Membership: |
|

DCTD
Genomic Map