Assay Details
| Gene Symbol: SEPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039854.1 | XP_017895343.1 | 9 - 10 | 960 | 68 | |
| GenBank mRNA | JO332572.2 | - | 4 - 5 | 888 | 68 | |
| JO633888.2 | - | 9 - 10 | 973 | 68 | ||
More Information
Set Membership: |
|

SEPT2
Genomic Map