Assay Details
| Gene Symbol: LOC102180106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963475.2 | XP_013818929.2 | 1 - 2 | 103 | 70 | |
| GenBank mRNA | JT688580.1 | - | 1 - 2 | 266 | 70 | |
More Information
Set Membership: |
|

LOC102180106
Genomic Map