Assay Details
| Gene Symbol: IRF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046944.1 | XP_017902433.1 | 5 - 6 | 656 | 63 | |
| XM_018046945.1 | XP_017902434.1 | 5 - 6 | 950 | 63 | ||
| XM_018046947.1 | XP_017902436.1 | 5 - 6 | 944 | 63 | ||
| XM_018046948.1 | XP_017902437.1 | 5 - 6 | 653 | 63 | ||
| XM_018046949.1 | XP_017902438.1 | 6 - 7 | 728 | 63 | ||
More Information
Set Membership: |
|

IRF5
Genomic Map