Assay Details
| Gene Symbol: ELANE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050498.1 | XP_017905987.1 | 4 - 5 | 647 | 95 | |
| GenBank mRNA | JO578546.1 | - | 3 - 4 | 513 | 95 | |
| JO578547.1 | - | 2 - 3 | 648 | 95 | ||
More Information
Set Membership: |
|

ELANE
Genomic Map