Assay Details
| Gene Symbol: LOC102172632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055300.1 | XP_017910789.1 | 10 - 11 | 1571 | 116 | |
| XM_018055301.1 | XP_017910790.1 | 10 - 11 | 1571 | 116 | ||
| XM_018055302.1 | XP_017910791.1 | 10 - 11 | 1571 | 116 | ||
More Information
Set Membership: |
|

LOC102172632
Genomic Map